Ethyl 3-bromo-1H-pyrazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A766893-1G A766893-250MG A766893-25G A766893-5G
77267-980EA 47.38 USD
77267-980 77267-982 77267-984 77267-986
Ethyl 3-bromo-1H-pyrazole-5-carboxylate 98%
Ethyl 3-bromo-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
MDL Number: MFCD28393486
CAS Number: 1886994-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR