3-(N,N-Dimethylamino)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(N,N-Dimethylamino)phenylboronic acid, pinacol ester, N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-(Dimethylamino)phenylboronic acid pinacol ester

A766561-100MG A766561-1G A766561-250MG
77281-246EA 35 USD
77281-246 77281-248 77281-250
3-(N,N-Dimethylamino)phenylboronic acid pinacol ester 98%
3-(N,N-Dimethylamino)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂
MDL Number: MFCD06795669
CAS Number: 325142-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR