2-Amino-3-methoxypropan-1-ol hydrochloride 97%

Supplier: Ambeed
Warning

A766519-100MG A766519-250MG
77267-962EA 34 USD
77267-962 77267-964
2-Amino-3-methoxypropan-1-ol hydrochloride 97%
2-Amino-3-methoxypropan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂ClNO₂
MDL Number: MFCD28125204
CAS Number: 333743-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR