4,7-Bis(5-bromothiophen-2-yl)-5,6-difluorobenzo[c][1,2,5]thiadiazole 98%

Supplier: Ambeed
Warning

A766408-100MG A766408-250MG
77243-970EA 66.95 USD
77243-970 77243-972
4,7-Bis(5-bromothiophen-2-yl)-5,6-difluorobenzo[c][1,2,5]thiadiazole 98%
4,7-Bis(5-bromothiophen-2-yl)-5,6-difluorobenzo[c][1,2,5]thiadiazole

Precaución: for Research Use Only

Formula: C₁₄H₄Br₂F₂N₂S₃
MW: 494.19 g/mol
MDL Number: MFCD30186157
CAS Number: 1304773-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR