N-(2-Iodophenyl)acetamide 98%

Supplier: Ambeed
Warning

A766307-1G A766307-250MG
77267-954EA 135.96 USD
77267-954 77267-956
N-(2-Iodophenyl)acetamide 98%
N-(2-Iodophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₈INO
MW: 261.05 g/mol
MDL Number: MFCD00099251
CAS Number: 19591-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR