(S)-2-((S)-2-Benzyl-4-((5-methyl-1H-imidazol-4-yl)methyl)-3-oxopiperazine-1-carboxamido)-4-methylpentanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: GGTI-2418

A765852-10MG A765852-1MG A765852-25MG A765852-50MG A765852-5MG A765852-100MG
77281-230EA 107 USD
77281-230 77281-232 77281-234 77281-236 77281-238 77749-523
(S)-2-((S)-2-Benzyl-4-((5-methyl-1H-imidazol-4-yl)methyl)-3-oxopiperazine-1-carboxamido)-4-methylpentanoic acid 98%
(S)-2-((S)-2-Benzyl-4-((5-methyl-1H-imidazol-4-yl)methyl)-3-oxopiperazine-1-carboxamido)-4-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₃₁N₅O₄
MW: 441.52 g/mol
MDL Number: MFCD25976818
CAS Number: 501010-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR