Nonaethylene glycol monododecylether (C12E9) 99%

Supplier: Ambeed
Warning

A765105-100G A765105-25G A765105-500G A765105-5G
77267-944EA 78 USD
77267-944 77267-946 77267-948 77267-950
Nonaethylene glycol monododecylether (C12E9) 99%
Nonaethylene glycol monododecylether (C12E9)

Precaución: for Research Use Only

Formula: C₃₀H₆₂O₁₀
MDL Number: MFCD00043375
CAS Number: 3055-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR