2,4-Diisopropylphenol 95%

Supplier: Ambeed
Warning

A764597-250MG A764597-1G A764597-5G A764597-25G
77255-698EA 53.01 USD
77255-698 77573-466 77747-512 77747-511
2,4-Diisopropylphenol 95%
2,4-Diisopropylphenol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
MDL Number: MFCD01707536
CAS Number: 2934-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR