N-(Benzo[d]thiazol-2-yl)-4-((2-hydroxy-3-methoxybenzyl)amino)benzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: ML355

A764546-10MG A764546-1MG A764546-25MG A764546-5MG
77243-958EA 203.94 USD
77243-958 77243-960 77243-962 77243-964
N-(Benzo[d]thiazol-2-yl)-4-((2-hydroxy-3-methoxybenzyl)amino)benzenesulfonamide 98%
N-(Benzo[d]thiazol-2-yl)-4-((2-hydroxy-3-methoxybenzyl)amino)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₃O₄S₂
MW: 441.52 g/mol
MDL Number: MFCD28167822
CAS Number: 1532593-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR