2-Methoxy-5-methylbenzonitrile 95%

Supplier: Ambeed
Warning

A764363-10G A764363-1G A764363-5G
77267-938EA 75 USD
77267-938 77267-940 77267-942
2-Methoxy-5-methylbenzonitrile 95%
2-Methoxy-5-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD03425693
CAS Number: 53078-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR