(2-Chlorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorobenzyl cyanide, 2-(2-Chlorophenyl)acetonitrile

A763761-1000G A763761-100G A763761-10G A763761-25G A763761-500G
77267-928EA 53.56 USD
77267-928 77267-930 77267-932 77267-934 77267-936
(2-Chlorophenyl)acetonitrile 98%
(2-Chlorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClN
MW: 151.6 g/mol
MDL Number: MFCD00001898
CAS Number: 2856-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR