Adamantane-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A763690-100MG A763690-1G A763690-250MG A763690-25G A763690-5G
77255-688EA 10.3 USD
77255-688 77255-690 77255-692 77255-694 77573-464
Adamantane-2-carboxylic acid 95%
Adamantane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.24 g/mol
MDL Number: MFCD02738135
CAS Number: 15897-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR