4-Benzyloxy-1-butanol 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Benzyloxy)butan-1-ol, 1-Butanol, 4-(phenylmethoxy)-, 4-benzyloxy-butan-1-ol, 4-benzyloxybutanol

A763682-10G A763682-1G A763682-5G
77293-590EA 103 USD
77293-590 77293-592 77293-594 77293-596
4-Benzyloxy-1-butanol 97%
4-Benzyloxy-1-butanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.247 g/mol
MDL Number: MFCD00155211
CAS Number: 4541-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR