2-Methyl-3-((2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-yl)naphthalene-1,4-dione 98%

Supplier: Ambeed
Warning

A763662-1MG A763662-1G A763662-250MG A763662-100MG
77281-228EA 7 USD
77281-228 77618-106 77618-108 77749-522
2-Methyl-3-((2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-yl)naphthalene-1,4-dione 98%
2-Methyl-3-((2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-yl)naphthalene-1,4-dione

Precaución: for Research Use Only

Formula: C₃₁H₄₀O₂
MDL Number: MFCD00079646
CAS Number: 863-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR