1,2,4-Trifluoro-5-nitrobenzene 98%

Supplier: Ambeed
Danger

A763036-10G A763036-25G A763036-5G
77293-584EA 8 USD
77293-584 77293-586 77293-588
1,2,4-Trifluoro-5-nitrobenzene 98%
1,2,4-Trifluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₃NO₂
MDL Number: MFCD00007051
CAS Number: 2105-61-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR