Acetonitrilebis[2-diphenylphosphino-6-t-butylpyridine]cyclopentadienylruthenium(II) hexafluorophosphate 98%

Supplier: Ambeed
Warning

A763016-100MG A763016-50MG
77255-684EA 315 USD
77255-684 77255-686
Acetonitrilebis[2-diphenylphosphino-6-t-butylpyridine]cyclopentadienylruthenium(II) hexafluorophosphate 98%
Acetonitrilebis[2-diphenylphosphino-6-t-butylpyridine]cyclopentadienylruthenium(II) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₄₉H₅₂F₆N₃P₃Ru
MW: 990.94 g/mol
MDL Number: MFCD06798306
CAS Number: 776230-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR