2,2'-Dithienyl disulfide 97%

Supplier: Ambeed
Danger

A762061-1G A762061-5G
77281-222EA 15 USD
77281-222 77281-224 77281-226
2,2'-Dithienyl disulfide 97%
2,2'-Dithienyl disulfide

Precaución: for Research Use Only

Formula: C₈H₆S₄
MDL Number: MFCD00066333
CAS Number: 6911-51-9
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR