[1,1'-Biphenyl]-3,4',5-tricarbaldehyde 98%

Supplier: Ambeed
Warning

A761846-100MG A761846-1G A761846-250MG
77267-916EA 21.63 USD
77267-916 77267-918 77267-920
[1,1'-Biphenyl]-3,4',5-tricarbaldehyde 98%
[1,1'-Biphenyl]-3,4',5-tricarbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₃
MW: 238.23 g/mol
MDL Number: MFCD31700812
CAS Number: 187281-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR