4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate 98%

Supplier: Ambeed
Warning

Synonyms: meso-Tetraphenylporphine-4,4′,4″,4″′-tetrasulphonic acid tetrasodium salt, 4,4',4'',4'''-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulphonic acid)tetrasodium salt xhydrate, 5,10,15,20-Tetraphenyl-21H, 23H-porphine-p,p′,p″,p′′′-tetrasulphonic acid tetrasodium hydrate

A761659-100MG A761659-1G A761659-250MG
77255-678EA 45 USD
77255-678 77255-680 77255-682
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate 98%
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate

Precaución: for Research Use Only

Formula: C₄₄H₂₆N₄Na₄O₁₂S₄·xH₂O
MDL Number: MFCD10567204
CAS Number: 652154-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR