3-Fluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluorobenzene-1-sulphonyl chloride

A760992-10G A760992-1G A760992-25G A760992-5G
77293-574EA 20.6 USD
77293-574 77293-576 77293-578 77293-580
3-Fluorobenzenesulfonyl chloride 98%
3-Fluorobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₄ClFO₂S
MW: 194.61 g/mol
MDL Number: MFCD00042286
CAS Number: 701-27-9
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR