3,6-Dichloro-1-benzothiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: BT2, 3,6-Dichlorobenzo[b]thiophene-2-carboxylic acid

A760956-100MG A760956-10G A760956-1G A760956-250MG A760956-5G A760956-25G
77281-212EA 9.27 USD
77281-212 77281-214 77281-216 77281-218 77281-220 77749-519
3,6-Dichloro-1-benzothiophene-2-carboxylic acid 97%
3,6-Dichloro-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₄Cl₂O₂S
MW: 247.1 g/mol
MDL Number: MFCD00781745
CAS Number: 34576-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR