2,2',7,7'-Tetrakis(diphenylamino)-9,9'-spirobi[9H-fluorene] 97%

Supplier: Ambeed
Warning

A760845-1G A760845-250MG A760845-5G
77255-672EA 58 USD
77255-672 77255-674 77255-676
2,2',7,7'-Tetrakis(diphenylamino)-9,9'-spirobi[9H-fluorene] 97%
2,2',7,7'-Tetrakis(diphenylamino)-9,9'-spirobi[9H-fluorene]

Precaución: for Research Use Only

Formula: C₇₃H₅₂N₄
MDL Number: MFCD11114061
CAS Number: 189363-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR