5-(2-Ethoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one methanesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sildenafil Mesylate
A760525-100MG
A760525-1MG
A760525-25MG
77267-910EA
153.47
USD
77267-910
77267-912
77267-914
5-(2-Ethoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one methanesulfonate 98%
5-(2-Ethoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one methanesulfonate
Precaución: for Research Use Only
Formula:
C₂₃H₃₄N₆O₇S₂ MW: 570.68 g/mol |
MDL Number:
MFCD09753069 CAS Number: 1308285-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...