4-Iodo-2-methyl-1-nitrobenzene 97%

Supplier: Ambeed
Danger

A759773-100MG A759773-1G A759773-250MG
77293-568EA 19.57 USD
77293-568 77293-570 77293-572
4-Iodo-2-methyl-1-nitrobenzene 97%
4-Iodo-2-methyl-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MW: 263.033 g/mol
MDL Number: MFCD08234827
CAS Number: 52415-00-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR