3,5-Di(2-pyridyl)pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(1H-Pyrazole-3,5-diyl)dipyridine

A759650-100MG A759650-250MG A759650-5G
77281-204EA 30.9 USD
77281-204 77281-206 77281-208
3,5-Di(2-pyridyl)pyrazole 98%
3,5-Di(2-pyridyl)pyrazole

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₄
MW: 222.25 g/mol
MDL Number: MFCD00130125
CAS Number: 129485-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR