2-Hydrazinylpyridine dihydrochloride 95%

Supplier: Ambeed
Warning

A759586-1G A759586-5G
77267-900EA 36 USD
77267-900 77267-902
2-Hydrazinylpyridine dihydrochloride 95%
2-Hydrazinylpyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₉Cl₂N₃
MDL Number: MFCD00012814
CAS Number: 62437-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR