1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-3,4-dihydronaphthalene-2-carbaldehyde

A759563-100MG A759563-1G A759563-250MG
77243-938EA 37.08 USD
77243-938 77243-940 77243-942
1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde 98%
1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₉ClO
MW: 192.65 g/mol
MDL Number: MFCD02257735
CAS Number: 3262-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR