4,4'-Methylenedibenzonitrile 95%

Supplier: Ambeed
Warning

A758867-10G A758867-250MG
77293-562EA 149 USD
77293-562 77293-564
4,4'-Methylenedibenzonitrile 95%
4,4'-Methylenedibenzonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₀N₂
MW: 218.25 g/mol
MDL Number: MFCD09032983
CAS Number: 10466-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR