Methyl 4-bromo-2,6-dichlorobenzoate 98%

Supplier: Ambeed
Warning

A758513-1G A758513-5G
77255-668EA 55 USD
77255-668 77573-446
Methyl 4-bromo-2,6-dichlorobenzoate 98%
Methyl 4-bromo-2,6-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O₂
MDL Number: MFCD28037387
CAS Number: 232275-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR