3-Mercaptopicolinic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: SKF-34288 HCl

A758333-10MG A758333-1MG A758333-5MG
77281-192EA 30.9 USD
77281-192 77281-194 77281-196
3-Mercaptopicolinic acid hydrochloride 95%
3-Mercaptopicolinic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MW: 191.635 g/mol
MDL Number: MFCD01320417
CAS Number: 320386-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR