4H-Cyclopenta[b]thiophen-6(5H)-one 98%

Supplier: Ambeed
Warning

A758241-250MG A758241-1G
77293-560EA 80.34 USD
77293-560 77749-513
4H-Cyclopenta[b]thiophen-6(5H)-one 98%
4H-Cyclopenta[b]thiophen-6(5H)-one

Precaución: for Research Use Only

Formula: C₇H₆OS
MW: 138.1869 g/mol
MDL Number: MFCD03426936
CAS Number: 5650-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR