N-Boc-(R)-2-Amino-3-methoxy-1-propanol 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl [(2R)-1-hydroxy-3-methoxypropan-2-yl]carbamate, (R)-tert-Butyl (1-hydroxy-3-methoxypropan-2-yl)carbamate

A757534-1G A757534-5G
77267-896EA 28.84 USD
77267-896 77267-898
N-Boc-(R)-2-Amino-3-methoxy-1-propanol 95%
N-Boc-(R)-2-Amino-3-methoxy-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₄
MW: 205.251 g/mol
MDL Number: MFCD20261174
CAS Number: 183793-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR