2-Methyl-4-trifluoromethyl-1H-imidazole 95%

Supplier: Ambeed
Warning

A757525-250MG A757525-1G A757525-5G
77281-178EA 22 USD
77281-178 77618-096 77749-512
2-Methyl-4-trifluoromethyl-1H-imidazole 95%
2-Methyl-4-trifluoromethyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂
MDL Number: MFCD10000863
CAS Number: 33468-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR