2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole 98%

Supplier: Ambeed
Warning

Synonyms: PBD, 2-([1,1-Biphenyl]-4-yl)-5-(4-(tert-butyl)phenyl)-1,3,4-oxadiazole, 2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole

A757355-25G A757355-5G
77243-918EA 150 USD
77243-918 77243-920
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole 98%
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₂O
MW: 354.45 g/mol
MDL Number: MFCD00003101
CAS Number: 15082-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR