3-Methyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A757202-100MG A757202-1G A757202-5G A757202-250MG
77281-174EA 18 USD
77281-174 77281-176 77618-092 77749-511
3-Methyl-9H-carbazole 98%
3-Methyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₃H₁₁N
MDL Number: MFCD00464338
CAS Number: 4630-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR