tert-Butyl 2-hydroxy-1H-benzo[d]imidazole-1-carboxylate 95%

Supplier: Ambeed
Warning

A757181-1G A757181-5G
77267-894EA 88.58 USD
77267-894 77618-090
tert-Butyl 2-hydroxy-1H-benzo[d]imidazole-1-carboxylate 95%
tert-Butyl 2-hydroxy-1H-benzo[d]imidazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃
MW: 234.25 g/mol
MDL Number: MFCD11226799
CAS Number: 161468-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR