4-Fluoro-3-nitrobenzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-Fluoro-3-nitrobenzenesulphonyl Chloride

A757078-1G A757078-250MG A757078-5G
77293-552EA 52.53 USD
77293-552 77293-554 77293-556
4-Fluoro-3-nitrobenzenesulfonyl chloride 95%
4-Fluoro-3-nitrobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₄S
MW: 239.6100 g/mol
MDL Number: MFCD06342770
CAS Number: 6668-56-0
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR