3-(Trifluoromethoxy)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A756379-1G A756379-25G A756379-5G
77281-154EA 14 USD
77281-154 77281-156 77281-158
3-(Trifluoromethoxy)benzenesulfonyl chloride 98%
3-(Trifluoromethoxy)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O₃S
MDL Number: MFCD01091016
CAS Number: 220227-84-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR