Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride 98%

Supplier: Ambeed
Warning

A756333-100MG A756333-1G A756333-250MG
77255-662EA 95 USD
77255-662 77255-664 77255-666
Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride 98%
Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride

Precaución: for Research Use Only

Formula: C₅₈H₅₄ClP₂RuCl
MW: 984.9900 g/mol
MDL Number: MFCD09753014
CAS Number: 131614-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR