N,N-Dimethyl-5-(piperazin-1-ylsulfonyl)naphthalen-1-amine 97%

Supplier: Ambeed
Warning

A756309-100MG A756309-1G A756309-250MG
77267-886EA 31 USD
77267-886 77267-888 77267-890
N,N-Dimethyl-5-(piperazin-1-ylsulfonyl)naphthalen-1-amine 97%
N,N-Dimethyl-5-(piperazin-1-ylsulfonyl)naphthalen-1-amine

Precaución: for Research Use Only

Formula: C₁₆H₂₁N₃O₂S
MDL Number: MFCD06411705
CAS Number: 86516-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR