3,3-Dimethylbutyric acid 98%

Supplier: Ambeed
Danger

A756265-10G A756265-25G
77293-548EA 6 USD
77293-548 77293-550
3,3-Dimethylbutyric acid 98%
3,3-Dimethylbutyric acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MDL Number: MFCD00002715
CAS Number: 1070-83-3
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR