2-Methyl-2-nitropropane-1,3-diol 98%

Supplier: Ambeed
Warning

A756221-100G A756221-1G A756221-25G A756221-5G
77267-872EA 284 USD
77267-872 77267-874 77267-876 77267-878
2-Methyl-2-nitropropane-1,3-diol 98%
2-Methyl-2-nitropropane-1,3-diol

Precaución: for Research Use Only

Formula: C₄H₉NO₄
MDL Number: MFCD00024812
CAS Number: 77-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR