3-Fluorobenzenesulfonamide 97%
Supplier: Ambeed
|
Warning
|
A755791-10G
A755791-1G
A755791-250MG
A755791-25G
A755791-5G
77293-538EA
40
USD
77293-538
77293-540
77293-542
77293-544
77293-546
3-Fluorobenzenesulfonamide 97%
3-Fluorobenzenesulphonamide
Precaución: for Research Use Only
Formula:
C₆H₆FNO₂S |
MDL Number:
MFCD00042284 CAS Number: 1524-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...