2,4-Dimethoxybenzenesulfonyl chloride 90%
Supplier: Ambeed
|
Danger
|
A755689-10G
A755689-1G
A755689-5G
77255-648EA
29
USD
77255-648
77255-650
77747-484
2,4-Dimethoxybenzenesulfonyl chloride 90%
2,4-Dimethoxybenzenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₈H₉ClO₄S |
MDL Number:
MFCD00060683 CAS Number: 63624-28-2 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...