Amino-PEG6-t-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 1-amino-3,6,9,12,15,18-hexaoxahenicosan-21-oate, NH2-PEG6-Boc

A754913-100MG A754913-250MG A754913-1G A754913-5G
77255-644EA 89.61 USD
77255-644 77255-646 77618-084 77618-086
Amino-PEG6-t-butyl ester 95%
Amino-PEG6-t-butyl ester

Precaución: for Research Use Only

Formula: C₁₉H₃₉NO₈
MW: 409.51 g/mol
MDL Number: MFCD13184944
CAS Number: 1286281-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR