2-Bromo-5-(piperidin-1-yl)pyridine 97%
Supplier: Ambeed
|
Warning
|
A754399-100MG
A754399-1G
A754399-250MG
77243-886EA
72.1
USD
77243-886
77243-888
77243-890
2-Bromo-5-(piperidin-1-yl)pyridine 97%
2-Bromo-5-(piperidin-1-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrN₂ MW: 241.12 g/mol |
MDL Number:
MFCD11869571 CAS Number: 1142197-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...