2,6-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A754235-100MG A754235-1G A754235-250MG A754235-50MG A754235-5G
77255-634EA 11 USD
77255-634 77255-636 77255-638 77255-640 77255-642
2,6-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((R)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₂₇N₃O₂
MDL Number: MFCD07368373
CAS Number: 185346-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR