tert-Butyl 4-(piperidin-3-ylmethyl)piperazine-1-carboxylate 97%
Supplier: Ambeed
|
Danger
|
A754079-100MG
A754079-1G
A754079-250MG
77243-880EA
52.53
USD
77243-880
77243-882
77243-884
tert-Butyl 4-(piperidin-3-ylmethyl)piperazine-1-carboxylate 97%
tert-Butyl 4-(piperidin-3-ylmethyl)piperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₂₉N₃O₂ MW: 283.40 g/mol |
MDL Number:
MFCD13248791 CAS Number: 1211569-09-3 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...