(E)-3-Benzylidenedihydrofuran-2(3H)-one 97%

Supplier: Ambeed
Warning

A752270-100MG A752270-1G A752270-250MG
77243-874EA 44 USD
77243-874 77243-876 77243-878
(E)-3-Benzylidenedihydrofuran-2(3H)-one 97%
(E)-3-Benzylidenedihydrofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
CAS Number: 30959-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR