2-Phenyl-2-propanol 98%

Supplier: Ambeed
Warning

Synonyms: α,α-Dimethylbenzyl alcohol

A751831-10G A751831-1G A751831-25G A751831-5G A751831-100G A751831-500G
77281-144EA 19.57 USD
77281-144 77281-146 77281-148 77552-298 77618-076 77749-509
2-Phenyl-2-propanol 98%
2-Phenyl-2-propanol

Precaución: for Research Use Only

Formula: C₆H₅C(OH)(CH₃)₂
MW: 136.19 g/mol
MDL Number: MFCD00004456
CAS Number: 617-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR